missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Dx Viral/Pathogen NA Isolation Kit

The Applied Biosystems MagMAX Dx Viral/Pathogen NA Isolation Kit is a magnetic-bead-based nucleic acid purification kit intended for the isolation and purification of viral and bacterial nucleic acid from respiratory biological specimens for downstream qPCR applications.

Brand:  Applied Biosystems™ A52736

841.50 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 17914771

  • 990.00 € / 600 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Applied Biosystems MagMAX Dx Viral/Pathogen NA Isolation Kit is a magnetic-bead-based nucleic acid purification kit intended for the isolation and purification of viral and bacterial nucleic acid from respiratory biological specimens for downstream qPCR applications.

Features of the MagMAX Dx Viral/Pathogen NA Isolation Kit include:
• Compatible with multiple downstream assays
• Single purification protocol for routinely tested pathogens and respiratory biological specimen without need for additional lysis
• Ability to purify various samples on single plate
• Sustainable product with biodegradable ingredients
• Designed to work with KingFisher Apex Dx instrument in under 40 mins

The kit is intended for use by qualified and trained clinical laboratory personnel specifically instructed and trained in the techniques of automated magnetic-bead purification and in vitro diagnostic procedures.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Binding Solution, 160 mL
• Wash Solution, 300 mL
• Binding Beads, 6 mL
• Proteinase K, 3 mL
• Elution Buffer, 36 mL

Shipped and stored at room temperature.

Magnetic Bead
KingFisher™ Apex Dx
Up to 600 preps
MagMAX™
Respiratory Pathogens
96-well Plate
Nasopharyngeal swab, Anterior nasal swab, Saliva, Sputum
Real-Time PCR
Viral and Bacterial
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For In Vitro Diagnostic Use