missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Dx Viral/Pathogen NA Isolation Kit

The Applied Biosystems MagMAX Dx Viral/Pathogen NA Isolation Kit is a magnetic-bead-based nucleic acid purification kit intended for the isolation and purification of viral and bacterial nucleic acid from respiratory biological specimens for downstream qPCR applications.

990.00 € - 3300.00 €

Specifications

Format 96-well Plate
Isolation Technology Magnetic Bead
Sample Type Nasopharyngeal swab, Anterior nasal swab, Saliva, Sputum
For Use With (Equipment) KingFisher™ Apex Dx
For Use With (Application) Real-Time PCR
View More Specs

Products 2
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
17914771
View Documents
Applied Biosystems™
A52736
Up to 600 preps
990.00 €
600 reactions
Please to purchase this item. Need a web account? Register with us today!
17934771
View Documents
Applied Biosystems™
A52735
Up to 2,000 preps
3300.00 €
2000 preps
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Applied Biosystems MagMAX Dx Viral/Pathogen NA Isolation Kit is a magnetic-bead-based nucleic acid purification kit intended for the isolation and purification of viral and bacterial nucleic acid from respiratory biological specimens for downstream qPCR applications.

Features of the MagMAX Dx Viral/Pathogen NA Isolation Kit include:
• For in vitro diagnostic use. This product is US-IVD and CE-IVD compliant.
• Single purification protocol for routinely tested respiratory pathogens without the need for additional lysis
• Ability to purify various sample types on a single plate
• Sustainable product with biodegradable ingredients and responsibly-sourced packaging
• Automation protocol that allows for 96 samples to be processed on the KingFisher Apex Dx Purification instrument with ∼30-minute run time

The kit is intended for use by qualified and trained clinical laboratory personnel specifically instructed and trained in the techniques of automated magnetic-bead purification and in vitro diagnostic procedures.

Instructions for Use approved for your registered country
Protocol for KingFisher Apex Dx instrument (Dx_MVP_RSP_APX_V1.kfdx)

Please check with your country specific representative for more information on availability.

 


Specifications

Specifications

96-well Plate
Nasopharyngeal swab, Anterior nasal swab, Saliva, Sputum
Real-Time PCR
MagMAX™
Respiratory Pathogens
Magnetic Bead
KingFisher™ Apex Dx
Viral and Bacterial
Room Temperature
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 24 Months

SDS
Documents

Documents

Product Certifications

For In Vitro Diagnostic Use