missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Chloride Hexahydrate, Fisher BioReagents
CAS: 7786-30-3 | Cl2H12MgO6 | 203.30 g/mol
Brand: Fisher BioReagents BP214-500
27.64 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
This item is not returnable.
View return policy
Description
Poly Bottle; 500g
Magnesium Chloride is suitable for use in the preparation of buffer solutions.
Specifications
Magnesium Chloride Hexahydrate | |
7786-30-3, 7791-18-6 | |
5 to 6.5 | |
500 g | |
≥99.0% | |
MFCD00011106 | |
magnesium chloride, mgcl2, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride | |
[Mg++].[Cl-].[Cl-] | |
203.30 | |
CHEBI:6636 | |
≥99.0% | |
>106°C |
117°C | |
White | |
Powder/Solid | |
≥99.0 % | |
Cl2H12MgO6 | |
15, 5726 | |
TWRXJAOTZQYOKJ-UHFFFAOYSA-L | |
magnesium;dichloride | |
5360315 | |
203.31 | |
Poly Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Magnesium Chloride Hexahydrate, Fisher BioReagents > Poly Bottle; 500g
Spot an opportunity for improvement?Share a Content Correction