Learn More
Buerkle™ Magnetic Stirring Bar
Molded with an outer layer of chemically inert PTFE for durability and non-reactivity. Buerkle™ Magnetic Stirring Bars feature a cylindrical shape and smooth surface for excellent mixing capability.
Brand:  Buerkle™ 8271-2507
 View more versions of this product
Product Code. 10184532
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Opaque PTFE construction
  • High performance permanent magnets 
  • Long lifespan
  • Exceptional temperature resistance
  • Wide range of sizes
  • Stirring bars with diameters of 6 and 8mm can be used with Buerkle™ Fish-clip™ for extra gentle stirring
Specifications
Specifications
White
7 mm
25 mm
Cylinder