Learn More
Buerkle™ Magnetic Stirring Bar
Molded with an outer layer of chemically inert PTFE for durability and non-reactivity. Buerkle™ Magnetic Stirring Bars feature a cylindrical shape and smooth surface for excellent mixing capability.
3.52 € - 14.95 €
Spezifikation
Color White
Shape Cylinder
Mehr Spezifikationen ansehen
Products 9
Produktcode Marke Length (Metric) Diameter (Metric) Preis Menge & Verfügbarkeit  
Produktcode Marke Length (Metric) Diameter (Metric) Preis Menge & Verfügbarkeit  
10581063
missing translation for 'viewDocuments' Promotion Details
Buerkle™
8271-2006
20 mm 6 mm
3.65 €
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
10676823
missing translation for 'viewDocuments' Promotion Details
Buerkle™
8271-2506
25 mm 6 mm
3.95 €
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
10143272
missing translation for 'viewDocuments' Promotion Details
Buerkle™
8271-3006
30 mm 6 mm
4.10 €
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
10184532
missing translation for 'viewDocuments' Promotion Details
Buerkle™
8271-2507
25 mm 7 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10011792
missing translation for 'viewDocuments' Promotion Details
Buerkle™
8271-3007
30 mm 7 mm
5.55 €
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
10051952
missing translation for 'viewDocuments' Promotion Details
Buerkle™
8271-4007
40 mm 7 mm
7.15 €
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
10134542
missing translation for 'viewDocuments' Promotion Details
Buerkle™
8271-4008
40 mm 8 mm
4.95 €
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
10581423
missing translation for 'viewDocuments' Promotion Details
Buerkle™
8271-5008
50 mm 8 mm
6.10 €
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
10451403
missing translation for 'viewDocuments' Promotion Details
Buerkle™
8271-8010
80 mm 10 mm
14.95 €
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Opaque PTFE construction
  • High performance permanent magnets 
  • Long lifespan
  • Exceptional temperature resistance
  • Wide range of sizes
  • Stirring bars with diameters of 6 and 8mm can be used with Buerkle™ Fish-clip™ for extra gentle stirring
Spezifikation
Spezifikation
White
Cylinder
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate