Learn More
Value Plastics Male Luer Slip Union Fittings
Luer fittings allow for a simple yet secure connection
Brand:  Value Plastics 50120-14
Product Code. 30053702
  • 24.14 € / Pack of 25
Estimated Shipment: 26-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Quick and easy assembly and disassembly without the need for tools. Constructed of durable chemical resistance nylon. Conforms to ISO-594-1 and ISO 594-2 standards
Specifications
Specifications
Non-DEHP;Non-Phthalate;RoHS
25/Pk.
Male Luer Slip
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Value Plastics Male Luer Slip Union Fittings > 25 Luer Fitting Union White Nylon ML Slip