missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MALP-2, TLR6 and TLR2 ligand

Applications: Functional, In vitro assay

Brand:  Novus Biologicals™ NBP2-26219

 View more versions of this product

Product Code. 18709023

  • 731.00 € / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specific Activity is approx. 2x10^8units/mg. One unit is defined as the dilution giving half maximal release of nitric oxide from C3H/HeJ mouse peritoneal exudate cells in the standard assay. Immunogen: MALP-2, TLR6 and TLR2 ligand Buffer: 2μg in 20μL of sterile PBS solution containing 2.5% (v/v) 2-propanol, 25mM n-octyl-b-D-glucopyranoside and 1% (wt/v) human serum albumin. Dilution: Functional, In vitro assay MALP-2 was originally isolated from Mycoplasma fermentans. This MALP-2 corresponds to the originally isolated isomer, which expresses potent endotoxin-like activity and approaches in certain experimental systems the toxicity of LPS. MW: 2135.2
Specifications

Specifications

0.1 mg/mL
Store at -20C. Avoid freeze-thaw cycles.
Source: Synthetic Formula: C99H167N19O30S MW: 2135.2 Use in In-Vitro and Functional Assay Reported in Scientific Literature (PMID: 25466255).
Human/Mouse/Avian
C99H167N19O30S
50 μg
MALP-2, TLR6 and TLR2 Ligand
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers