missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MALP-2, TLR6 and TLR2 ligand
Applications: Functional, In vitro assay
731.00 €
Specifications
Concentration | 0.1 mg/mL |
---|---|
Formulation | C99H167N19O30S |
Content And Storage | Store at -20C. Avoid freeze-thaw cycles. |
Target | MALP-2, TLR6 and TLR2 Ligand |
Species | Human/Mouse/Avian |
Description
Specific Activity is approx. 2x10^8units/mg. One unit is defined as the dilution giving half maximal release of nitric oxide from C3H/HeJ mouse peritoneal exudate cells in the standard assay. Immunogen: MALP-2, TLR6 and TLR2 ligand Buffer: 2μg in 20μL of sterile PBS solution containing 2.5% (v/v) 2-propanol, 25mM n-octyl-b-D-glucopyranoside and 1% (wt/v) human serum albumin. Dilution: Functional, In vitro assay MALP-2 was originally isolated from Mycoplasma fermentans. This MALP-2 corresponds to the originally isolated isomer, which expresses potent endotoxin-like activity and approaches in certain experimental systems the toxicity of LPS. MW: 2135.2Specifications
0.1 mg/mL | |
Store at -20C. Avoid freeze-thaw cycles. | |
Human/Mouse/Avian |
C99H167N19O30S | |
MALP-2, TLR6 and TLR2 Ligand |