missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
Packaging | Glass Bottle |
Quantity | 250 mg |
Certificate of Analysis | Available |
Physical Form | Powder |
Merck Index | 13-5738 |
Color | Yellow to Gray |
Chemical Name or Material | Mancozeb |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0459826 | Certificate of Analysis | 05/02/2024 | 468850010,468852500 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mancozeb, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction