Learn More
Manganese(II) chloride, 99+%, anhydrous, -80 mesh, Thermo Scientific Chemicals
47.13 € - 148.74 €
Chemical Identifiers
CAS | 7773-01-5 |
---|---|
Molecular Formula | Cl2Mn |
Molecular Weight (g/mol) | 125.84 |
MDL Number | MFCD00011114 |
InChI Key | GLFNIEUTAYBVOC-UHFFFAOYSA-L |
Synonym | manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline |
PubChem CID | 10313134 |
ChEBI | CHEBI:63041 |
IUPAC Name | manganese(2+) dichloride |
SMILES | [Cl-].[Cl-].[Mn++] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10495175
|
Thermo Scientific Acros
223610100 |
10 g | Glass bottle |
47.13 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10607382
|
Thermo Scientific Acros
223610500 |
50 g | Glass bottle |
148.74 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7773-01-5 | |
125.84 | |
GLFNIEUTAYBVOC-UHFFFAOYSA-L | |
10313134 | |
manganese(2+) dichloride |
Cl2Mn | |
MFCD00011114 | |
manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline | |
CHEBI:63041 | |
[Cl-].[Cl-].[Mn++] |
Specifications
1190°C | |
7773-01-5 | |
Powder | |
Cl2Mn | |
MFCD00011114 | |
15, 5793 | |
Solubility in water: 723g/L (25°C). Other solubilities: soluble in alcohol,insoluble in ether | |
[Cl-].[Cl-].[Mn++] | |
125.84 | |
CHEBI:63041 | |
≥99% |
650°C | |
Pink | |
99% min. (Complexometry) | |
MnCl2 | |
07,222 | |
manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline | |
GLFNIEUTAYBVOC-UHFFFAOYSA-L | |
manganese(2+) dichloride | |
10313134 | |
125.84 | |
Manganese(II) chloride, 99+% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Causes serious eye damage.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not eat,drink or smoke when using th
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-869-6
missing translation for 'rtecsNumber' : OO9625000
missing translation for 'tsca' : TSCA
- Manganese(II) chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Manganese(II) chloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.