missing translation for 'onlineSavingsMsg'
Learn More

Manganese(III) acetate hydrate, 96%, Thermo Scientific Chemicals

87.74 € - 377.88 €

Chemical Identifiers

CAS 19513-05-4
Molecular Formula C6H9MnO6
Molecular Weight (g/mol) 232.07
MDL Number MFCD00150022
InChI Key AHSBSUVHXDIAEY-UHFFFAOYSA-K
IUPAC Name manganese(3+) triacetate
SMILES [Mn+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11377288
View Documents
Thermo Scientific Alfa Aesar
030519.09
10 g
87.74 €
10g
Please to purchase this item. Need a web account? Register with us today!
11387288
View Documents
Thermo Scientific Alfa Aesar
030519.18
50 g
377.88 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It acts as a mild and selective oxidizing agent in organic synthesis and materials science. Manganese(III) Acetate Dihydrate is also a reagent used for radical cyclizations and α-keto-acetoxylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

19513-05-4
232.07
AHSBSUVHXDIAEY-UHFFFAOYSA-K
[Mn+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
C6H9MnO6
MFCD00150022
manganese(3+) triacetate

Specifications

19513-05-4
Crystalline
Mn(OOCCH3)3·xH2O
It is soluble in water.
[Mn+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
232.07
96%
1.59 g/mL
Pink
C6H9MnO6
MFCD00150022
AHSBSUVHXDIAEY-UHFFFAOYSA-K
manganese(3+) triacetate
232.07 (Anhydrous)
Hygroscopic
Manganese(III) acetate hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P501c

H315-H319-H335-H336-H373

missing translation for 'einecsNumber' : 213-602-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Manganese(III) acetate hydrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

Supplemental information
  • MIXTURE LIST-Contains : Manganese(III) acetate hydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only