missing translation for 'onlineSavingsMsg'
Learn More

Manganese(III) phosphate hydrate, 99%, Thermo Scientific Chemicals

32.00 € - 79.60 €

Chemical Identifiers

CAS 104663-56-1
Molecular Formula MnO4P
Molecular Weight (g/mol) 149.91
MDL Number MFCD00209658
InChI Key BECVLEVEVXAFSH-UHFFFAOYSA-K
PubChem CID 71317435
SMILES [Mn+3].[O-]P([O-])([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11403478
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A17868.09
10 g
32.00 €
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11413478
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A17868.18
50 g
79.60 €
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Insoluble in water.

Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

104663-56-1
149.91
BECVLEVEVXAFSH-UHFFFAOYSA-K
[Mn+3].[O-]P([O-])([O-])=O
MnO4P
MFCD00209658
71317435

Specifications

104663-56-1
MFCD00209658
BECVLEVEVXAFSH-UHFFFAOYSA-K
manganese(3+);phosphate;hydrate
71317435
99%
MnO4P
Insoluble in water.
[Mn+3].[O-]P([O-])([O-])=O
149.91
149.91 (Anhydrous)
Manganese(III) phosphate hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only