missing translation for 'onlineSavingsMsg'
Learn More

Mannitol, Powder, Multi-Compendial, U.S.P., J.T. Baker™

366.00 € - 818.00 €

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
PubChem CID 6251
ChEBI CHEBI:16899
SMILES OCC(O)C(O)C(O)C(O)CO
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15351791
View Documents
Avantor J.T.Baker
2553-01
500 g Poly Bottle
366.00 €
500g
Please to purchase this item. Need a web account? Register with us today!
15538024
View Documents
Avantor J.T.Baker
2553-05
2.5 kg Poly Bottle
818.00 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

69-65-8
182.17
FBPFZTCFMRRESA-UHFFFAOYNA-N
6251
OCC(O)C(O)C(O)C(O)CO
C6H14O6
MFCD00064287
d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
CHEBI:16899

Specifications

69-65-8
MFCD00064287
d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
OCC(O)C(O)C(O)C(O)CO
182.17
CHEBI:16899
Multi-Compendial/USP
Mannitol
C6H14O6
1L = 1.52kg
FBPFZTCFMRRESA-UHFFFAOYNA-N
(2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
6251
182.17
Powder
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.