Learn More
Fisherbrand™ Manual Decapper, Steel, Non-autoclavable
Manual decrimping tools allow easy removal of aluminum seals without breakage.
Brand:  Fisherbrand™ 08 07 0001
 View more versions of this product
Product Code. 11758276
  • 341.00 € / Each
Explore more special offers
Description
Description
  • Decapper
  • Manual
  • Steel
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Manual
Manual Decapper
Aluminum Cap
8 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Manual Decapper, Steel, Non-autoclavable > For 8mm caps