missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces

Reduce the amount of reagents used in the lab by using these plates and modules with covalent binding surfaces.

Brand:  Thermo Scientific™ 436007

921.12 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.


960.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10432612

  • 969.60 € / Pack of 30
Estimated Shipment: 02-02-2026
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Reduce the amount of reagents used in the lab with Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces.

Covalent Surfaces (Thermo Scientific™ CovaLink™ and Immobilizer Amino)

  • Minimal leaching
  • Withstands vigorous washing
  • Coating with lower amounts of reagent may be possible
  • Control of orientation

Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
8 x 12
Covalent
Solid plate 96-Well
400 μL
100 μL
Covalent Binding
Round, Flat Bottom (F-well)
5
Peptides
Chemiluminescence
30 Cases
White
Polystyrene
96
Non-sterile
400 μL
30
Unspecified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number176791 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number436007
Lot Number168479 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number436007
Lot Number177658 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number436007
Lot Number179064 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number436007
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces > Nunc Immobilizer, Solid plate 96-well, Flat-bottom, Amino, 400μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.