missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Manual Decapper, Steel, Non-autoclavable
Manual decrimping tools allow easy removal of aluminum seals without breakage.
Brand: Fisherbrand™ 11 07 0002
Description
- Decapper
- Manual
- Steel

Specifications
Manual | |
Manual Decapper | |
Aluminum Cap | |
11 mm |
Product Suggestions
Customers who viewed this item also viewed.
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Manual Decapper, Steel, Non-autoclavable > For 11mm caps
Spot an opportunity for improvement?Share a Content Correction