missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Watson-Marlow Tubing Marprene Process Tubing
This Marprene Process Tubing is made from a long life thermoplastic elastomer with resistance to a wide range of chemicals.
Brand: Watson-Marlow Tubing 902.0032.J16
Description
- Marprene is a long life thermoplastic elastomer with resistance to a wide range of chemicals.
- It lasts at least 10 times as long as tubing in other materials.
- Long-life tubing means stoppages for a tube change are rare events, easily fitted in with general maintenance.
- Marprene has wide chemical compatibility, and is highly resistant to oxidizing agents such as ozone, peroxides and sodium hypochlorite.
- Marprene is beige and opaque to visible and ultraviolet light, with low permeability to oxygen, carbon dioxide and nitrogen.
- It meets USDA standards for food handling.

Specifications
Autoclavable | |
3.2 mm | |
16 ft. | |
0.0625 in. | |
Beige | |
176°F | |
Process Tubing |
0.125 in. | |
TPE | |
5 m | |
1.6 mm | |
62 to 68 (Shore A) | |
80°C |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Watson-Marlow Tubing Marprene Process Tubing > Marprene Process Tubing; Inner Diameter: 3.2 mm
Spot an opportunity for improvement?Share a Content Correction