missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Mastercycler™ X40
GREENER_CHOICE

Mastercycler X40 combines properties that raise your standard in end-point PCR. Classic features such as a 12-column gradient and ideal block temperature homogeneity are enhanced by an easy, intuitive user interface, a touchscreen and a protective SafeLid to protect your samples from evaporation.

Brand:  Eppendorf™ 6381000042

 View more versions of this product

Product Code. 17889058

  • 6997.88 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • SafeLid for evaporation protection
  • Small footprint (less than a piece of paper)
  • Lightweight
  • Touchscreen
  • Intuitive software and user interface
  • Program migration feature
  • Visionize Labsuite connection possible
  • Low noise level
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fixed
20°C to 72°C ≤±0.2°C, 72°C to 95°C ≤±0.3°C
0.1 to 0.2 mL/Well
2 Years
Touchscreen
5000
37°C to 110°C
1.5°C/sec.
96-well
±0.15°C
24.5 x 38.5 x 23 cm (9.7 x 15.2 x 9.1 in.)
Thermal Cycler
4°C to 99°C
Aluminum
1°C to 30°C
3.3 °C/sec.
7.25 kg
UK
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eppendorf™ Mastercycler™ X40 > Mastercycler X40; Plug Type: UK

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.