missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Nylon Barbed Fittings

Hose barb fittings make tubing connections quick and easy, requiring no tools for installation.

Brand:  Masterflex™ 30613-26

 View more versions of this product

Product Code. 12344628

  • 32.85 € / Pack of 10
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Easily connect fluid paths in the laboratory
  • No leaks, flow restrictions or costly downtime
  • Creates seals that typically hold pressures up to 30 psi (2 bar)
Specifications

Specifications

Barbed Fitting
0.625 in. Tubing I.D.
10 Pack
T-shape
Nylon
-46°C
Ethylene Oxide
121°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.