missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ Nylon Barbed Fittings
Hose barb fittings make tubing connections quick and easy, requiring no tools for installation.
Brand: Masterflex™ 30616-18
This item is not returnable.
View return policy
Description
- Easily connect fluid paths in the laboratory
- No leaks, flow restrictions or costly downtime
- Creates seals that typically hold pressures up to 30 psi (2 bar)
Specifications
Barbed Fitting | |
0.375 x 0.25 in. Tubing I.D. | |
10 Pack | |
Straight |
Nylon | |
-46°C | |
Ethylene Oxide | |
121°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction