Learn More
Masterflex™ Single-Channel Polycarbonate Housing Standard Pump Head
Use the pump heads for common fluid transfer applications requiring accuracy and excellent repeatability. MasterFlex™ Single-channel Polycarbonate Housing Standard Pump Heads are perfect for space-limited applications and easy to integrate into your OEM applications.
274.00 € - 902.00 €
Specifications
No. of Channels | 1 |
---|---|
Occlusion Type | Fixed |
Quantity | 1 Each |
Type | Standard Pump Head |
Case Material | Polycarbonate |
Product Code | Brand | Flow Rate | Rotor Material | Tubing Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Flow Rate | Rotor Material | Tubing Size | Price | Quantity & Availability | ||||
11785158
|
Masterflex™
07013-20 |
0.06 to 36 mL/min. | Cold-rolled Steel | 13 |
274.00 €
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
11705168
|
Masterflex™
07016-20 |
0.8 to 480 mL/min. | Cold-rolled Steel | 16 |
274.00 €
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
11735168
|
Masterflex™
07018-20 |
3.8 to 2300 mL/min. | Cold-rolled Steel | 18 |
274.00 €
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
11800090
|
Masterflex™
07024-20 |
2.8 to 1700 mL/min. | Cold-rolled Steel | 24 |
274.00 €
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
11791199
|
Masterflex™
07035-20 |
3.8 to 2300 mL/min. | Cold-rolled Steel | 35 |
274.00 €
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
11770299
|
Masterflex™
07036-30 |
4.8 to 2900 mL/min. | Cold-rolled Steel | 36 |
364.00 €
Each |
Estimated Shipment: 19-06-2025
Log in to see stock. |
||||
11795059
|
Masterflex™
07019-20 |
0.002 to 8 L/min. | Cold-rolled Steel | 73 |
672.00 €
Each |
Estimated Shipment: 19-06-2025
Log in to see stock. |
||||
11755148
|
Masterflex™
07019-32 |
0.002 to 8 L/min. | Cold-rolled Steel | 82 |
708.00 €
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
11765089
|
Masterflex™
MFLX07013-21 |
0.06 to 36 mL/min. | Stainless Steel | 13 |
386.00 €
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
11795089
|
Masterflex™
07015-21 |
1.7 to 1000 mL/min. | Stainless Steel | 15 |
487.00 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
11775089
|
Masterflex™
07014-21 |
0.21 to 130 mL/min. | Stainless Steel | 14 |
386.00 €
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
11811455
|
Masterflex™
07019-21 |
0.002 to 8 L/min. | Stainless Steel | 19 |
902.00 €
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
11810090
|
Masterflex™
07024-21 |
2.8 to 1700 mL/min. | Stainless Steel | 24 |
386.00 €
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
11715099
|
Masterflex™
MFLX07018-21 |
3.8 to 2300 mL/min. | Stainless Steel | 18 |
386.00 €
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
Description
Use the pump heads for common fluid transfer applications requiring accuracy and excellent repeatability. MasterFlex™ Single-channel Polycarbonate Housing Standard Pump Heads are perfect for space-limited applications and easy to integrate into your OEM applications.
- High-precision economical pump head for precision tubing
- Standard heads are precision molded to accommodate a single size of tubing
- Available with clear polycarbonate (PC) housing with either cold-rolled steel (CRS) or stainless steel (SS) rotors
- CRS offers economy, while SS offers better chemical resistance and durability
- The 52 models with opaque black polyphenylene sulfide (PPS) housing and SS rotors offer the best overall chemical resistance
- Pump head is compact and simple in design with same side tubing entry/exit
- Standard heads are compact and easy to integrate into your OEM applications
Specifications
1 | |
1 Each | |
Polycarbonate |
Fixed | |
Standard Pump Head |