Learn More
Thermo Scientific™ Matrix™ Reagent Reservoirs

Accommodate a variety of applications with these uniquely designed, innovative reagent reservoirs.
Brand: Thermo Scientific™ 8085
112.00 EUR valid until 2025-05-31
Use promo code "25076" to get your promotional price.
Description
Accomodate a variety of applications with uniquely designed, innovative Thermo Scientific™ Matrix Reagent Reservoirs. These sterile reservoirs offer an extra-wide base to add rigidity and stability, helping to avoid spills. The 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizing recovery if using only a single channel and a small amount of reagent.
Features:
- Trough within a trough maximizes amount of liquid accessible to pipette tips when using small amounts of reagent
- Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
- Graduations on inside wall enable quick measurement of remaining liquid
- Sterile reservoirs are packaged in snap-and-tear bags, maintaining sterility while being easy to open

Specifications
White | |
Polystyrene | |
Reagent Reservoir |
100 mL | |
Sterile | |
10 Bags of 10 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.