Description
Ideal for cytotoxic and cytostatic waste which, to meet regulations, should be disposed of separately from other healthcare waste. The SHARPSGUARD™ cyto lid assists safe, easy collection of items including giving sets, syringes and gloves. Color-coded with a purple lid, handle and label, it can be safely transported for incineration after closure. Absorbent pads are included. The container is made to BS EN ISO 23907.Specifications
7 L |
Product Content Correction
Product Title
Polypropylene Disposal Bins > Capacity: 7L
Spot an opportunity for improvement?Share a Content Correction