Learn More
Fisherbrand™ Maxi Pipet Tips
GREENER_CHOICE
For use with large volume pipets
Brand:  Fisherbrand™ 090-FIS
 View more versions of this product
Product Code. 11512823
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
For use with large volume pipets
  • Designed for use when transferring large sample volume
  • Advanced molding technology ensure tips are straight
  • Alternative option to serological pipets
  • Nonsterile
  • Certified by independent laboratory to be RNase, DNase, DNA, and Pyrogen free
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1000 to 5000 μL
Non-sterile
Autoclavable
117 mm
Macro
No
Non-filtered
Natural
4.6 in.
Bulk
Gilson Pipetman
Polypropylene