missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Maxi Pipet Tips
GREENER_CHOICE

For use with large volume pipets

23.60 € - 93.00 €

Specifications

Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Format Bulk
Material Polypropylene
View More Specs

Products 7
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
11917754
GREENER_CHOICE
View Documents
Fisherbrand™
090-FIS
1000 to 5000 μL
32.65 €
Pack of 250
In Stock
to see stock.
Edge
Add to basket
 
11512823
GREENER_CHOICE
View Documents
Fisherbrand™
090-FIS
1000 to 5000 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17650627
View Documents
Fisherbrand™
02707467
1 to 5 mL
93.00 €
Pack of 250
In Stock
to see stock.
Edge
Add to basket
 
11937754
GREENER_CHOICE
View Documents
Fisherbrand™
095-FIS
1 to 5 mL
51.50 €
Pack of 250
In Stock
to see stock.
Edge
Add to basket
 
11997744
GREENER_CHOICE
View Documents
Fisherbrand™
089-FIS
500 to 2500 μL
38.58 €
Pack of 500
In Stock
to see stock.
Edge
Add to basket
 
11648138
GREENER_CHOICE
View Documents
Fisherbrand™
096-FIS
5000 μL
23.60 €
Pack of 250
In Stock
to see stock.
Edge
Add to basket
 
11987744
GREENER_CHOICE
View Documents
Fisherbrand™
085-FIS
250 to 2500 μL
29.80 €
Pack of 500
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
 
Description

Description

For use with large volume pipets

  • Designed for use when transferring large sample volume
  • Advanced molding technology ensure tips are straight
  • Alternative option to serological pipets
  • Nonsterile
  • Certified by independent laboratory to be RNase, DNase, DNA, and Pyrogen free
Specifications

Specifications

Non-sterile
Autoclavable
Polypropylene
Natural
Bulk
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.