missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MaxQ™ Shaker Universal Platforms

Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms.

Brand:  Thermo Scientific™ 30110

 View more versions of this product

Product Code. 10798914

  • 483.41 € / Each
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms. With exceptional innovation, reliability and accuracy, these platforms are the optimal choice for a busy lab.

For use with MaxQ Shakers as indicated. Choose the platform that best suits your needs.

Ordering Information:

Purchase clamps separately.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Universal Platform
18 x 18 in. (45.7 x 45.7 cm)
45.7 cm
45.7 cm
MaxQ™ 2000, 2506, 2508, 4000, 6000 Shaker
Certificate of Analysis (COA), Certificate of Conformance (COC)
18 in.
18 in.
Universal Platform without Clamps, 18 x 18 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ MaxQ™ Shaker Universal Platforms > Universal platform without clamps; 18 x 18 in.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.