Learn More
Axygen™ Maxymum Recovery™ 200 μL Universal Pipetter Tips: 200 μL, Beveled
Designed to fit a wide variety of single and multi-channel pipettes
Brand:  Axygen™ T-200-C-L
24.28 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 12655445
  • 32.23 € / Pack of 1000
Estimated Shipment: 04-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Maxymum Recovery technology for ultra-low retention
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 to 200 μL
Clear
DNase, RNase, DNA, and PCR inhibitor Free
Beveled, Low Retention
Universal Fit
Polypropylene
Non-sterile
Autoclavable
Bulk
Bulk; 1,000 Tips/Pk, 20 Packs/Cs
Maxymum Recovery
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Axygen™ Maxymum Recovery™ 200 μL Universal Pipetter Tips: 200 μL, Beveled > Bulk; Maxymum Recovery; 1-200 μL