missing translation for 'onlineSavingsMsg'
Learn More

MBL™ PTFE Stopcock Modular Class B Burette

Manufactured from borosilicate glass and with funnel top to make filling easier and safer

Brand:  MBL BRB264P

33.68 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12441567

  • 36.35 € / Each
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Complies with ISO 385
  • With funnel top to make filling easier and safer
  • PTFE stopcock for grease-free performance
  • Easy to read blue enamel graduations
  • Modular design with detachable burette tube, stopcock and jet for easy replacement
TRUSTED_SUSTAINABILITY
Specifications

Specifications

25 mL
0.1 mL
PTFE
Volumetric Buret
Borosilicate Glass
Blue
±0.1 mL
ISO 385
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.