missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ MDCC, 7-Diethylamino-3-((((2-Maleimidyl)ethyl)amino)carbonyl)coumarin
7-diethylamino-3-((((2-maleimidyl)ethyl)amino)carbonyl)coumarin (MDCC)
Brand: Molecular Probes™ D10253
This item is not returnable.
View return policy
Description
The thiol reactive coumarin, MDCC can be used to create blue-fluorescent bioconjugates. When compared with AMCA conjugates, conjugates of the UV light-excitable 7-dialkylaminocoumarin fluorophore have slightly longer-wavelength emission spectra (∼470 nm).
Specifications
5 mg | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
Methylcoumarin | |
470 |
MDCC | |
Thiol | |
Maleimide | |
Classic Dyes |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Molecular Probes™ MDCC, 7-Diethylamino-3-((((2-Maleimidyl)ethyl)amino)carbonyl)coumarin > 5mg
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction