missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Measuring Jug Industrial with Closed Handle

Light and handy polypropylene measuring jug is suitable for industrial applications.

Brand:  Buerkle™ 7204-5000

24.94 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 16606563

  • 25.80 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The measuring jug has two measuring scales, embossed transparent and with blue lettering. With ergonomic handle, available in closed design for special stability or with open handle for stacking to save space.The measuring jug is made of polypropylene and is therefore highly resistant to acids, bases and chemicals.
  • PP, transparent
  • With embossed graduation printed in blue
Specifications

Specifications

Polypropylene
5000 mL
257 mm
203 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buerkle™ Measuring Jug Industrial with Closed Handle > 5000 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.