Learn More
TFA Dostmann Mechanical Stopwatch
This mechanical stopwatch made of metal is particularly sturdy and always ready for use. The stopwatch is wound up by hand and then runs for up to 30 minutes. After the stop, you can read the time to 1/5 second. Carry bag and neck strap are included.
Brand:  TFA Dostmann 38.1022
Product Code. 16331278
  • 95.65 € / Each
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
This mechanical stopwatch made of metal is particularly sturdy and always ready for use. It is traditionally used for timekeeping in training or competition, but it can also be used in many other areas, such as industry, crafts and laboratories. The stopwatch is wound up by hand and then runs for up to 30 minutes. After the stop, you can read the time to 1/5 second. Carry bag and neck strap are included.
  • Start/stop with crown
  • Lateral reset push button
  • Time-out possible
  • Wind-up mechanism
  • 13 jewels
  • Chromed metal case
  • 1/5 second increments
  • Including neck strap and bag
Specifications
Specifications
Analog Stopclock
0 to 30 min.
98 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
TFA Dostmann Mechanical Stopwatch > Analog Stopclock