Learn More
Corning™ Medium 199 (Mod.)
Guaranteed low endotoxin levels ≤0.025 EU/mL
Brand: Corning™ 10-060-CV
This item is not returnable.
View return policy
Description
- Complete line of media and reagents for mammalian in vitro cell culture
- Ready-to-use media is supplied in polyethylene teraphthalate (PET) plastic bottles
- Sonically welded cap liners eliminate toxic resins or glues that could leach into media
- Shipped at ambient temperature
Specifications
2° to 8°C | |
Medium 199 | |
L-Glutamine, Earleâ€TMs Salts | |
7 to 7.4 |
1 X | |
<0.25EU/ mL | |
To assess growth promotion and cytotoxicity characteristics | |
6/Pk. |