missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Medium 199 (Mod.)
BIOPHARMA

Guaranteed low endotoxin levels ≤0.025 EU/mL

151.98 €

Specifications

Content And Storage 2° to 8°C
pH 7 to 7.4
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15343541
BIOPHARMA
View Documents
Corning™
10-060-CV
6/Pk.
151.98 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Complete line of media and reagents for mammalian in vitro cell culture
  • Ready-to-use media is supplied in polyethylene teraphthalate (PET) plastic bottles
  • Sonically welded cap liners eliminate toxic resins or glues that could leach into media
  • Shipped at ambient temperature

Specifications

Specifications

2° to 8°C
7 to 7.4
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 18 Months

SDS
Documents

Documents

Product Certifications