Learn More
OHAUS™ Medium Two Prong Sleeves
OHAUS™ Medium Two Prong Sleeves are for use with clamps and can be easily removed for cleaning or replacement.
10.25 €
Specifications
Quantity 2
Type Prong Sleeve
Size Medium
View More Specs
Products 2
Product Code Brand Material Price Quantity & Availability  
Product Code Brand Material Price Quantity & Availability  
15722669
View Documents
OHAUS™
30400253
Vinyl
10.25 €
Each
In Stock
to see stock.
Add to basket
15732669
View Documents
OHAUS™
30400254
Fiberglass
10.25 €
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Available in red vinyl and fiberglass
  • Fiberglass sleeves are ideal for application above 100°C
Specifications
Specifications
2
Prong Sleeve
Medium
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
OHAUS™ Medium Two Prong Sleeves