missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Melatonine, 99% Product Code.: 10316960

Thermo Scientific Chemicals Melatonine, 99%

Product Code. 10316960
1 g, Glass bottle
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10316960

Brand: Thermo Scientific Chemicals 125360010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Melatonine, 99%, CAS # 73-31-4, is a hormone usually produced in the pineal gland and released into the blood.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Melatonine is a hormone produced by the pineal gland that shows several biological activities, including the somnolence, sleep-wake cycle, circadian rhythm, stress response, aging, and immunity activities

• It shows antioxidant, autocoid, paracoid, hormonal, anti- inflammatory and anti-apoptotic activities

Applications

• Melatonin shows neurohormonal activity and can upregulate antioxidant enzymes and inhibit rat NOS1

• Melatonin has been reported to be an activator of MEL-1A-R and MEL-1B-R

• The production and secretion of melatonine are dependent on beta-adrenergic receptor function

• During melatonine synthesis, the essential amino acid L-tryptophan is used as a precursor

• This compound inhibits apoptosis in immune cells and neurons, and enhances apoptotic cell death of cancer cells

• It can inhibit proliferation and metastasis of breast cancer cells by modulating Rho-associated kinase protein-1 expression

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-31-4
Molecular Formula C13H16N2O2
Molecular Weight (g/mol) 232.28
MDL Number MFCD00005655
InChI Key DRLFMBDRBRZALE-UHFFFAOYSA-N
Synonym melatonin, melatonine, n-acetyl-5-methoxytryptamine, circadin, 5-methoxy-n-acetyltryptamine, n-2-5-methoxy-1h-indol-3-yl ethyl acetamide, melatol, melatonex, melovineShow MoreShow Less
PubChem CID 896
ChEBI CHEBI:16796
IUPAC Name N-[2-(5-methoxy-1H-indol-3-yl)ethyl]acetamide
SMILES CC(=O)NCCC1=CNC2=C1C=C(C=C2)OC

Specifications

Melting Point 116.0°C to 120.0°C
Color Cream to Yellow or White
Loss on Drying 0.5% max. (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 22, V,12, 42
Merck Index 15, 5884
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,benzene,toluene,hexane,acet,onitrile
Formula Weight 232.28
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Melatonine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461766 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number125360010,125360050,125362500
Lot NumberA0456000 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number125360010,125360050,125362500
Lot NumberA0451195 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number125360010,125360050,125362500
Lot NumberA0403804 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number125360010,125360050,125362500
Lot NumberA0396427 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number125360010,125360050,125362500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Melatonine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.