Learn More
Gibco™ MEM Amino Acids Solution (50X)
GREENER_CHOICE
MEM Amino Acids Solution (50X)
Brand:  Gibco™ 11130077
 View more versions of this product
Product Code. 11580386
  • 518.00 € / Pack of 20
Explore more special offers
Description
Description
Gibco™ MEM Amino Acids Solution is used as a growth supplement for cell culture medium, to increase cell growth and viability. Gibco™ MEM Amino Acids Solution is formulated to contain 50X the essential amino acids (except L-glutamine) found in the standard Minimum Essential Medium (MEM). The complete formulation is available.
Product Use
For in vitro diagnostic use.
Dual-site cGMP Manufacturing and Quality System
Gibco™ MEM Amino Acids Solution is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ MEM Amino Acids Solution product made in our Grand Island facility (11130-051). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 13485 standard, FDA
MEM Amino Acids Solution, 20 x 100mL, Liquid, 50X Concentration, Without Glutamine, Store at 2 to 8°C, Ambient Shipping Condition, 12 Months Shelf Life, For Mammalian Cell Culture
Liquid
No Glutamine
Mammalian Cell Culture
For in-vitro diagnostic use
2 to 8°C
50 X
Amino Acid Supplement
L-Arginine hydrochloride, L-cystine, L-histidine hydrochloride-H2O, L-isoleucine, L-leucine, L-lysine hydrochloride, L-methionine, L-phenylalanine, L-threonine, L-tryptophan, L-tyrosine, L-valine
20 x 100 mL