missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM Non-Essential Amino Acids Solution (100X)
GREENER_CHOICE

MEM Non-Essential Amino Acids Solution (100X)

23.17 € - 348.00 €

Specifications

Certifications/Compliance FDA, ISO 13485
Content And Storage 2 to 8°C
Concentration 100 X
Product Type Amino Acid Supplement
For Use With (Application) Mammalian Cell Culture
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12084947
GREENER_CHOICE
View Documents
Gibco™
11140035
100 mL
23.17 €
100mL
Please to purchase this item. Need a web account? Register with us today!
12070346
GREENER_CHOICE
View Documents
Gibco™
11140068
20 x 100 mL
348.00 €
Pack of 20
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gibco™ MEM Non-Essential Amino Acids Solution is used as a growth supplement for cell culture medium, to increase cell growth and viability. Gibco™ MEM Non-Essential Amino Acids Solution is formulated to contain 100X the non-essential amino acids found in the standard Minimum Essential Medium (MEM). The complete formulation is available.

Product Use
For in vitro diagnostic use.

Dual-site cGMP Manufacturing and Quality System
Gibco™ MEM Non-Essential Amino Acids Solution is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ MEM Non-Essential Amino Acids Solution product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications

Specifications

FDA, ISO 13485
100 X
Mammalian Cell Culture
2 to 8°C
Amino Acid Supplement
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 12 Months

Product Identifier
  • MEM NON-ESSENTIAL AMINO ACIDS SOLUTION, (100X)

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : amino acids

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.