missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Ion Selective Membrane Replacement Kits

Sample preparation steps such as filtering and distillation are rarely necessary with this kit for the combined ion-sensitive electrode with a solid membrane.

Brand:  Fisherbrand™ 11769728

 View more versions of this product

Product Code. 11769728

  • 351.00 € / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket

Description

Description

  • ORP electrode Fisherbrand 1m cable with BNC epoxy body 12mm x 120mm
  • ORP Electrode with BNC Epoxy Body
  • Combined ion-sensitive electrode with solid membrane interchangeable flat surface
  • The ion to be detected passes through the membrane generating a difference of potential proportional to ionic concentration
  • Analysis time generally less than one minute
  • Sample preparation steps such as filtering or distillation rarely necessary
  • Calibration should be carried out as often as possible (less once a day) at the same temperature as the sample
  • Measurement temperature 0 to 50 ° C
Specifications

Specifications

ion Selective Membrane Replacement Kits
FOR 83925
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers