missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Menzel™ Microscope Coverslips

Square microscope coverslips constructed of borosilicate glass.

Brand:  Epredia™ BB02400240A153MNZ0

 View more versions of this product

Product Code. 17294904

  • 29.45 € / Pack of 1000
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Square
  • Thickness of 0.13 to 0.16 mm
Specifications

Specifications

Coverslip
Borosilicate Glass
24 x 24 mm
Square
#1 (0.13 to 0.16 mm)
Automated Equipment
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers