Learn More
Mercury(II) nitrate hydrate, ACS, 98.0% min, Thermo Scientific Chemicals
52.42 € - 234.00 €
Chemical Identifiers
CAS | 7783-34-8 |
---|---|
Molecular Formula | H2HgN2O7 |
Molecular Weight (g/mol) | 342.62 |
MDL Number | MFCD00149736 |
InChI Key | KVICROHOONHSRH-UHFFFAOYSA-N |
Synonym | mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2 |
PubChem CID | 3084029 |
IUPAC Name | mercury(2+) hydrate dinitrate |
SMILES | O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Description
Mercury(II) nitrate is used as an oxidizing agent involved in organic synthesis. Its reaction with acetone yielded an organometallic compound containing mercury. In addition, it serves as a nitrification agent and analytical reagents used in the laboratory. It is involved in the preparation of mercury fulminate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7783-34-8 | |
342.62 | |
KVICROHOONHSRH-UHFFFAOYSA-N | |
3084029 | |
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
H2HgN2O7 | |
MFCD00149736 | |
mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2 | |
mercury(2+) hydrate dinitrate |
Specifications
79°C | |
White | |
H2HgN2O7 | |
MFCD00149736 | |
14,5880 | |
Soluble in water. | |
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
342.62 | |
324.60 (Anhydrous) | |
ACS | |
4.39 g/mL |
7783-34-8 | |
Crystalline | |
Hg(NO3)2·xH2O (x=1 to 2) | |
UN1625 | |
mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2 | |
KVICROHOONHSRH-UHFFFAOYSA-N | |
mercury(2+) hydrate dinitrate | |
3084029 | |
≤98.0% | |
Hygroscopic | |
Mercury(II) nitrate hydrate |
Safety and Handling
GHS H Statement
H300-H310-H330-H272-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May intensify fire
oxidizer.
May cause damage to organs through prolonged or repeated exposure.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
H300+H310+H330-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURIC NITRATE
missing translation for 'einecsNumber' : 233-152-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.