Learn More
Mercury(II) silver iodide, Thermo Scientific™
51.50 € - 466.00 €
Chemical Identifiers
CAS | 7784-03-4 |
---|---|
Molecular Formula | Ag2HgI4 |
Molecular Weight (g/mol) | 923.94 |
MDL Number | MFCD00046160 |
InChI Key | YNTJRFXPYRRRKS-UHFFFAOYSA-J |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7784-03-4 | |
923.94 | |
YNTJRFXPYRRRKS-UHFFFAOYSA-J |
Ag2HgI4 | |
MFCD00046160 |
Specifications
7784-03-4 | |
Powder | |
MFCD00046160 | |
YNTJRFXPYRRRKS-UHFFFAOYSA-J | |
923.95 | |
Mercury(II) silver iodide |
Yellow | |
Ag2HgI4 | |
UN2025 | |
923.94 | |
Light sensitive |
Safety and Handling
GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
H300+H310+H330-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.
missing translation for 'einecsNumber' : 232-045-9
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.