missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) telluride, 99.999% (metals basis), Thermo Scientific Chemicals

134.67 € - 557.78 €

Chemical Identifiers

CAS 12068-90-5
Molecular Formula HgTe
Molecular Weight (g/mol) 328.19
MDL Number MFCD00016133
InChI Key VCEXCCILEWFFBG-UHFFFAOYSA-N
Synonym mercury telluride, mercury ii telluride, mercuric telluride, quecksilbertellurid, mercurous telluride, mercury ii telluride trace metals basis 5g
PubChem CID 82914
IUPAC Name tellanylidenemercury
SMILES [Te]=[Hg]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11364177
View Documents
Thermo Scientific Alfa Aesar
036666.04
2 g
134.67 €
2g
Please to purchase this item. Need a web account? Register with us today!
11374177
View Documents
Thermo Scientific Alfa Aesar
036666.09
10 g
557.78 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Mercury cadmium telluride (MCT)is the mostim- portantmaterial currently in use for the manufacture of high performance infrared (IR) detectors. Mercury telluride and mercury telluride-cadmium telluride are semiconductors which have been investigated as intrinsic infrared detecting materials. Complete coverage of the low-loss fiber windows by mercury telluride quantum dot are used in colloidal nanocrystals for telecommunications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12068-90-5
328.19
VCEXCCILEWFFBG-UHFFFAOYSA-N
82914
[Te]=[Hg]
HgTe
MFCD00016133
mercury telluride, mercury ii telluride, mercuric telluride, quecksilbertellurid, mercurous telluride, mercury ii telluride trace metals basis 5g
tellanylidenemercury

Specifications

673°C
Black to Gray
HgTe
UN2025
Insoluble in water.
[Te]=[Hg]
328.19
328.19
Odorless
(metals basis)
12068-90-5
Fused Lump
MFCD00016133
mercury telluride, mercury ii telluride, mercuric telluride, quecksilbertellurid, mercurous telluride, mercury ii telluride trace metals basis 5g
VCEXCCILEWFFBG-UHFFFAOYSA-N
tellanylidenemercury
82914
99.999%
8.63 g/mL
Mercury(II) telluride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c

H300+H310+H330-H373

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.

missing translation for 'einecsNumber' : 235-108-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.