Learn More
Gibco™ MesenPRO RS™ Medium

MesenPRO RS™ Medium
Brand: Gibco™ 12746012
230.85 EUR valid until 2025-12-31
Use promo code "24342" to get your promotional price.
Description
MesenPRO RS™ Medium is a reduced serum (2%) medium specifically formulated to support the growth of human mesenchymal stem cells (MSCs) in culture. MesenPRO RS™ Medium consistently improves MSC expansion compared with conventional serum-supplemented media. Using MesenPRO RS™ Medium, MSCs can be expanded for multiple passages while maintaining their multipotential phenotype (i.e., differentiation into osteogenic, chondrogenic, and adipogenic lineages).
MesenPRO RS™ Medium consists of:- Basal liquid medium (MesenPRO RS™ Basal Medium)
- Frozen supplement (MesenPRO RS™ Growth Supplement), which is added at the time of use.
Formulated for the growth and expansion of mesenchymal stem cells.
Note: MesenPRO RS™ Medium has been developed for culturing MSCs at greater than clonal densities.
Recommended storage conditions:
- MesenPRO RS™ Basal Medium: Store in the dark at 2 to 8°C
- MesenPRO RS™ Growth Supplement: Store in the dark at -5 to -20°C
Both the Basal Medium and Growth Supplement are guaranteed stable for 24 months when properly stored.
Note: MesenPRO RS™ complete medium (basal, supplement and glutamine) is stable for 15 days when stored in the dark at 2 to 8°C. We do not recommend using the complete medium beyond 15 days.
Intended use(s):
in vitro diagnostic (IVD)
This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.

Specifications
Mesenchymal Stem Cells | |
Liquid | |
Reduced Serum | |
MesenPRO™ | |
Human | |
Adherent Cell Culture |
Serum-supplemented | |
Stem Cell Media | |
No Glutamine, No Phenol Red | |
1 Kit | |
MesenPRO RS™ Medium consists of: • 500 ml MesenPRO RS™ Basal Medium (store in the dark at 2°C to 8°C) • 10 ml MesenPRO RS™ Growth Supplement (store in the dark at -5°C to -20°C) The Basal Medium ships at room temperature, the Growth Supplement on dry ice. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.