missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Essential 6™ Medium
Essential 6™ Medium
Brand: Gibco™ A1516401
This item is not returnable.
View return policy
Description
Essential 6 Medium is a feeder-free and xeno-free medium that supports the reprogramming of somatic cells and the spontaneous or directed differentiation of human pluripotent stem cells (PSCs). In addition, Essential 6 Medium can be used as a base for custom media for the culture of PSCs. The formulation is based on a medium originally developed by Guokai Chen et. al. (1) in the laboratory of James Thomson and published as “E6.” With only six essential components, Essential 6 Medium helps minimize variability. To complete your workflow with a matching reduced-variability PSC culture medium developed by the same lab, use Essential 8 Medium.Essential 6 Medium enables:
- Differentiation—does not contain bFGF, which inhibits differentiation
- Reprogramming—does not contain TGFβ, which has a negative effect on reprogramming efficiency
- Flexibility—provides a flexible format where the levels of TGFβ and bFGF can be adjusted and additional components can be added to match a given application
Unlike other media used in PSC culture, Essential 6 Medium does not contain bFGF or TGFβ. As such, Essential 6 Medium can support the formation of embryoid bodies. It has also been used as a base for the directed differentiation of various cell types in the endodermal, mesodermal, and ectodermal lineages (2), including motor neurons (3).
Reprogramming
Essential 6 Medium allows for defined and feeder-free reprogramming when used with bFGF. The formulation supports somatic cell reprogramming using a variety of methods, including episomal vectors (4) and CytoTune (Sendai virus), and is optimized to help ensure maximum cell health and pluripotency with minimal variability.
Flexibility
Essential 6 Medium is xeno-free and contains only the essential components needed for stem cell culture, minus bFGF and TGFβ. This provides a basal medium that will maximize cell health and pluripotency while allowing levels of TGFβ and bFGF to be adjusted and additional components to be added to match a given application.
Commercialized in partnership with Cellular Dynamics International.

Specifications
Pluripotent Stem Cells | |
Liquid | |
Serum-free | |
cGMP for medical devices, 21 CFR Part 820 and ISO 13485 | |
500 mL | |
Human | |
Stem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic) |
Xeno-free | |
Stem Cell Media | |
Phenol Red | |
Essential 6™ | |
Ambient | |
Store at 2 to 8°C and protect from light. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Modulating Growth Factor Addition to Essential 6 Media for a Complete Xeno-free Media
File size: 2.3 MB
Essential media system for reprogramming
File size: 1.3 MB
Brochure: Gibco Essential 8 Media
File size: 652.8 kB
Essential 6 Medium
Version: Rev 2.0
File size: 127.4 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Essential 6™ Medium > 500mL
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction