missing translation for 'onlineSavingsMsg'
Learn More

meso-Hydrobenzoin, 95%, Thermo Scientific Chemicals

76.38 € - 180.90 €

Chemical Identifiers

CAS 579-43-1
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00064253
InChI Key IHPDTPWNFBQHEB-UHFFFAOYNA-N
Synonym meso-hydrobenzoin, meso-1,2-diphenyl-1,2-ethanediol, unii-co9a49a84i, meso-stilbene glycol, 1r,2s-1,2-diphenylethane-1,2-diol, meso-1,2-diphenylethylene glycol, hydrobenzoin, meso, unii-q61g3433lb component
PubChem CID 853018
ChEBI CHEBI:50015
IUPAC Name (1R,2S)-1,2-diphenylethane-1,2-diol
SMILES OC(C(O)C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11486816
View Documents
Thermo Scientific Alfa Aesar
B23946.06
5 g
76.38 €
5g
Please to purchase this item. Need a web account? Register with us today!
11496816
View Documents
Thermo Scientific Alfa Aesar
B23946.14
25 g
180.90 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

meso- Hydrobenzoin has been used in the preparation of trans-methyl meso- hydrobenzoin phosphite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
meso- Hydrobenzoin has been used in the preparation of trans-methyl meso- hydrobenzoin phosphite.

Solubility
Soluble in hot alcohol or chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Specifications

Chemical Identifiers

579-43-1
214.26
IHPDTPWNFBQHEB-UHFFFAOYNA-N
853018
(1R,2S)-1,2-diphenylethane-1,2-diol
C14H14O2
MFCD00064253
meso-hydrobenzoin, meso-1,2-diphenyl-1,2-ethanediol, unii-co9a49a84i, meso-stilbene glycol, 1r,2s-1,2-diphenylethane-1,2-diol, meso-1,2-diphenylethylene glycol, hydrobenzoin, meso, unii-q61g3433lb component
CHEBI:50015
OC(C(O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

579-43-1
C14H14O2
2050814
meso-hydrobenzoin, meso-1,2-diphenyl-1,2-ethanediol, unii-co9a49a84i, meso-stilbene glycol, 1r,2s-1,2-diphenylethane-1,2-diol, meso-1,2-diphenylethylene glycol, hydrobenzoin, meso, unii-q61g3433lb component
IHPDTPWNFBQHEB-UHFFFAOYNA-N
(1R,2S)-1,2-diphenylethane-1,2-diol
853018
214.27
meso-Hydrobenzoin
134°C to 136°C
MFCD00064253
14,4777
Soluble in hot alcohol or chloroform.
OC(C(O)C1=CC=CC=C1)C1=CC=CC=C1
214.26
CHEBI:50015
99%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 207-758-3

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.