missing translation for 'onlineSavingsMsg'
Learn More
meso-Tetraphenylporphine, low chlorine, Thermo Scientific Chemicals Product Code.: 11453807

meso-Tetraphenylporphine, low chlorine, Thermo Scientific Chemicals

Product Code. 11453807
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11453807

Brand: Thermo Scientific Alfa Aesar A18261.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

meso-Tetraphenylporphyrin is a porphyrin-based compound of interest in molecular electronics and supramolecular building blocks. Porphyrins are often used to construct structures in supramolecular chemistry. It finds it application as a sensitizer used for the photochemical generation of singlet oxygen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
meso-Tetraphenylporphyrin is a porphyrin-based compound of interest in molecular electronics and supramolecular building blocks. Porphyrins are often used to construct structures in supramolecular chemistry. It finds it application as a sensitizer used for the photochemical generation of singlet oxygen.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 917-23-7
Molecular Formula C44H30N4
Molecular Weight (g/mol) 614.75
MDL Number MFCD00011680
InChI Key AQPPOLXYUQPDOD-UHFFFAOYSA-N
Synonym tetraphenylporphyrin, 5,10,15,20-tetraphenylporphyrin, meso-tetraphenylporphine, tetraphenylporphine, 5,10,15,20-tetraphenylporphine, meso-tetraphenylporphyrin, 21h,23h-porphine, 5,10,15,20-tetraphenyl, 5,10,15,20-tetraphenyl-21h,23h-porphine, meso-tetraphenylporphyrine, tetraphenyl porphineShow MoreShow Less
PubChem CID 70186
SMILES N1C2=CC=C1C(=C1C=CC(=N1)C(=C1C=CC(=N1)C(=C1NC(C=C1)=C2C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 449°C to 451°C
Quantity 1 g
Beilstein 379542
Solubility Information Insoluble in water.
IUPAC Name 5,10,15,20-tetraphenyl-21,22-dihydroporphyrin
Formula Weight 614.71
Chemical Name or Material meso-Tetraphenylporphine, low chlorine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10186200 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18261.03,A18261.14,A18261.06
Lot Number5004A08N Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18261.03,A18261.14,A18261.06
Lot Number10189986 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18261.03,A18261.14,A18261.06
Lot Number10220285 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18261.03,A18261.14,A18261.06
Lot Number10159273 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18261.03,A18261.14,A18261.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
meso-Tetraphenylporphine, low chlorine, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.