Learn More
Nickel Electro™ Metalware Range™ Stainless Steel Square Pipet Sterilizing Boxes
Keep pipets sterile with a sterilizing box. Nickel Electro™ Metalware Range™ Stainless Steel Square Pipet Sterilizing Boxes can be steam sterilized at 120°C and withstand dry heat temperatures of up to 180°C.
Brand:  Nickel Electro™ 6706/SS
 View more versions of this product
Product Code. 12996986
  • 67.00 € / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stainless steel square profile
  • Includes lid
  • Square shape prevents it from rolling off bench top
  • Silicon in lid and base
Specifications
Specifications
Autoclavable
Stainless Steel
Each
Sterilizing Box
400 x 70 mm
Sterile
400 mm
Autoclave