missing translation for 'onlineSavingsMsg'
Learn More
Methanesulfonic acid, 70% aq. soln., Thermo Scientific Chemicals Product Code.: 11413398

Methanesulfonic acid, 70% aq. soln., Thermo Scientific Chemicals

Product Code. 11413398
500 mL
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy

Product Code. 11413398

Brand: Thermo Scientific Alfa Aesar A17803.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methane sulfonic acid 70% solution is used virtually in all electronic applications involving tin or tin-alloy plating from non fluoborate systems. It replaces phosphoric acid in detergent formulations, thereby helping to avoid environmental phosphate pollution. It is used in the preparation of polyaniline/graphene nanocomposites, which enhances its thermal and electrical properties. It is utilized for electropolishing of aluminum as well as in electroplating of noble metals like gold, platinum, palladium and their alloys.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methane sulfonic acid 70% solution is used virtually in all electronic applications involving tin or tin-alloy plating from non fluoborate systems. It replaces phosphoric acid in detergent formulations, thereby helping to avoid environmental phosphate pollution. It is used in the preparation of polyaniline/graphene nanocomposites, which enhances its thermal and electrical properties. It is utilized for electropolishing of aluminum as well as in electroplating of noble metals like gold, platinum, palladium and their alloys.

Solubility
Miscible with water. Slightly miscible with benzene and toluene. Immiscible with paraffins.

Notes
Light and heat sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with amines, strong reducing agents, ethyl vinyl ether, hydrofluoric acid, strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-75-2
Molecular Formula CH4O3S
Molecular Weight (g/mol) 96.1
MDL Number MFCD00007518
InChI Key AFVFQIVMOAPDHO-UHFFFAOYSA-N
Synonym methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethaneShow MoreShow Less
PubChem CID 6395
ChEBI CHEBI:27376
IUPAC Name methanesulfonic acid
SMILES CS(=O)(=O)O

Specifications

Density 1.32
Flash Point >110°C (230°F)
Refractive Index 1.415
Quantity 500 mL
UN Number UN2586
Merck Index 14,5954
Solubility Information Miscible with water. Slightly miscible with benzene and toluene. Immiscible with paraffins.
Formula Weight 96.1
Concentration or Composition (by Analyte or Components) 70% aq. soln.
Chemical Name or Material Methanesulfonic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462092 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA17803.AE,A17803.AP,A17803.0F
Lot NumberA0456978 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog NumberA17803.AE,A17803.AP,A17803.0F
Lot NumberA9880A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17803.AE,A17803.AP,A17803.0F
Lot Number10225952 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17803.AE,A17803.AP,A17803.0F
Lot Number10196451 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17803.AE,A17803.AP,A17803.0F
compliance-icons compliance-icons
Product Identifier
  • Methanesulphonic acid, 70% aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Methanesulphonic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methanesulfonic acid, 70% aq. soln., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.