missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, Laboratory Reagent, ≥99.6%, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
10 L
20 L
25 L
4 x 2.5 L
4 x 4 L
4 x 5 L
160 kg
Packaging:
Fiberboard Box
Glass Bottle
Metal Drum
Stainless Steel Can
Description
- ≥95% for general laboratory use.

Specifications
Specifications
Melting Point | -98°C |
Color | Colorless |
Density | 0.791g/mL at 25°C |
Boiling Point | 65°C |
Quantity | 10 L |
Explosion Limit | 0.36 |
Flash Point | 9°C (48.2°F) |
Ignition Point | 725°F |
Packaging | Stainless Steel Can |
Physical Form | Liquid |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 13
Safety and Handling



Product Identifier
- Methanol
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Methanol, Laboratory Reagent, ≥99.6%, Honeywell™ >
Spot an opportunity for improvement?Share a Content Correction