missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, Optima™ LC/MS Grade, Fisher Chemical
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Description
Methanol, >-99.9%, CH4O, CAS Number-67-56-1, wood alcohol, methyl hydroxide, colonial spirit, columbian spirit, wood naphtha, carbinol, methyl alcohol, pyroxylic spirit, wood spirit, methylol

Specifications
Specifications
Melting Point | -98°C |
Absorbance | ≤0.005 AU at 260nm, ≤0.005 AU at 280nm, ≤0.01 AU at 254nm, ≤0.1 AU at 230nm, ≤0.2 AU at 220nm, ≤0.4 AU at 214nm, ≤0.5 AU at 210nm |
Color | Colorless |
Density | 0.791g/cm³ |
Boiling Point | 64.7°C |
Quantity | 500 mL |
Identification | Pass Test |
Packaging | Amber Glass |
Physical Form | Liquid |
Assay Percent Range | Assay: 99.9% min by GC |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2525766 | Certificate of Analysis | 10/02/2025 | A456-500,A456-212,A456-1,A456-4 |
2524396 | Certificate of Analysis | 10/02/2025 | A456-500,A456-212,A456-1,A456-4 |
2524573 | Certificate of Analysis | 10/02/2025 | A456-500,A456-212,A456-1,A456-4 |
2524826 | Certificate of Analysis | 10/02/2025 | A456-500,A456-212,A456-1,A456-4 |
2525611 | Certificate of Analysis | 10/02/2025 | A456-500,A456-212,A456-1,A456-4 |
Safety and Handling



Product Identifier
- Methanol
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P240-Ground/bond container and receiving equipment.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Methanol, Optima™ LC/MS Grade, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction